Anti-BOB1 Mouse Monoclonal Antibody [clone: ABT-BOB1]

Supplier: ANTIBODIES.COM LLC
A99846-50 A99846-100
77080-206EA 350.2 USD
77080-206 77080-290
Anti-BOB1 Mouse Monoclonal Antibody [clone: ABT-BOB1]
Antibodies
Mouse monoclonal (ABT-BOB1) antibody to BOB.1 for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: BOB1
Clonality: Monoclonal
Clone: ABT-BOB1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR