Anti-S100 alpha 6 Mouse Monoclonal Antibody [clone: ABT-S100A6]

Supplier: ANTIBODIES.COM LLC
A98937-100 A98937-50
77062-992EA 484.2 USD
77062-992 77065-254
Anti-S100 alpha 6 Mouse Monoclonal Antibody [clone: ABT-S100A6]
Antibodies
Mouse monoclonal (ABT-S100A6) antibody to S100A6 for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: S100 alpha 6
Clonality: Monoclonal
Clone: ABT-S100A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR