Anti-Thyroglobulin Mouse Monoclonal Antibody [clone: ABT-TG]

Supplier: ANTIBODIES.COM LLC
A98339-100 A98339-50
77062-890EA 484.2 USD
77062-890 77080-474
Anti-Thyroglobulin Mouse Monoclonal Antibody [clone: ABT-TG]
Antibodies
Mouse monoclonal (ABT-TG) antibody to Thyroglobulin for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: Thyroglobulin
Clonality: Monoclonal
Clone: ABT-TG
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR