Anti-MSH6 Mouse Monoclonal Antibody [clone: ABT-MSH6]

Supplier: ANTIBODIES.COM LLC
A94716-50 A94716-100
77047-410EA 356.8 USD
77047-410 77047-408
Anti-MSH6 Mouse Monoclonal Antibody [clone: ABT-MSH6]
Antibodies
Mouse monoclonal (ABT-MSH6) antibody to MutS Protein Homolog 6 for IHC and WB with samples derived from Human.

  • Validated Applications: IHC, WB

Type: Primary
Antigen: MSH6
Clonality: Monoclonal
Clone: ABT-MSH6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR