[(4S)-4,5-Dihydro-4-phenyl-2-oxazolyl]ferrocene ≥98%, ee 99%

Supplier: Ambeed
Warning

A999089-100MG A999089-1G A999089-250MG
77257-028EA 9 USD
77257-028 77257-030 77257-032
[(4S)-4,5-Dihydro-4-phenyl-2-oxazolyl]ferrocene ≥98%, ee 99%
[(4S)-4,5-Dihydro-4-phenyl-2-oxazolyl]ferrocene

Precaución: for Research Use Only

Formula: C₁₉H₁₇FeNO
MW: 331.18 g/mol
MDL Number: MFCD30475626
CAS Number: 162157-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR