3-(Trimethylsilylethynyl)thiophene 98%

Supplier: Ambeed
Warning

Synonyms: Trimethyl(thiophen-3-ylethynyl)silane

A998427-1G A998427-250MG A998427-5G
77282-150EA 21.63 USD
77282-150 77282-152 77282-154
3-(Trimethylsilylethynyl)thiophene 98%
3-(Trimethylsilylethynyl)thiophene

Precaución: for Research Use Only

Formula: C₉H₁₂SSi
MW: 180.34 g/mol
MDL Number: MFCD04039959
CAS Number: 130995-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR