(R)-3,3'-Bis(9-phenanthrenyl)-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate 98%

Supplier: Ambeed
Warning

A996076-100MG A996076-1G A996076-250MG
77257-012EA 85 USD
77257-012 77257-014 77257-016
(R)-3,3'-Bis(9-phenanthrenyl)-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate 98%
(R)-3,3'-Bis(9-phenanthrenyl)-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate

Precaución: for Research Use Only

Formula: C₄₈H₂₉O₄P
MDL Number: MFCD12546016
CAS Number: 864943-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR