1-Oxaspiro[2.5]octane-2-carbonitrile 98+%

Supplier: Ambeed
Danger

A995641-1G A995641-250MG
77245-026EA 55.62 USD
77245-026 77245-028
1-Oxaspiro[2.5]octane-2-carbonitrile 98+%
1-Oxaspiro[2.5]octane-2-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MDL Number: MFCD00101211
CAS Number: 36929-66-5
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR