5,7-Dibromobenzo[d]isoxazol-3-amine 97%

Supplier: Ambeed
Warning

A995340-100MG A995340-250MG
77269-174EA 64 USD
77269-174 77269-176
5,7-Dibromobenzo[d]isoxazol-3-amine 97%
5,7-Dibromobenzo[d]isoxazol-3-amine

Precaución: for Research Use Only

Formula: C₇H₄Br₂N₂O
MDL Number: MFCD29762366
CAS Number: 89583-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR