3-Ethoxy-2-nitropyridine 98+%

Supplier: Ambeed
Warning

A995059-1G A995059-250MG A995059-5G
77282-138EA 43.26 USD
77282-138 77282-140 77282-142
3-Ethoxy-2-nitropyridine 98+%
3-Ethoxy-2-nitropyridine

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
MDL Number: MFCD00006257
CAS Number: 74037-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR