R-3,3'-Dibromo-1,1'-binaphthyl-2,2'-diylhydrogenphosphate 98%
Supplier: Ambeed
|
Warning
|
A991527-100MG
A991527-250MG
A991527-5G
A991527-1G
77257-008EA
120.51
USD
77257-008
77257-010
77747-743
77747-742
R-3,3'-Dibromo-1,1'-binaphthyl-2,2'-diylhydrogenphosphate 98%
R-3,3'-Dibromo-1,1'-binaphthyl-2,2'-diylhydrogenphosphate
Precaución: for Research Use Only
Formula:
C₂₀H₁₁Br₂O₄P MW: 506.08 g/mol |
CAS Number:
861909-33-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...