Methyl 4-formyl-2-hydroxybenzoate 95%

Supplier: Ambeed
Warning

A991196-100MG A991196-1G A991196-250MG
77269-166EA 124.63 USD
77269-166 77269-168 77269-170
Methyl 4-formyl-2-hydroxybenzoate 95%
Methyl 4-formyl-2-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈O₄
MW: 180.16 g/mol
MDL Number: MFCD24674004
CAS Number: 42137-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR