trans-4-(Cyclopropylmethoxy)cyclohexanamine 97%

Supplier: Ambeed
Warning

A991135-100MG A991135-1G A991135-250MG
77269-160EA 39.14 USD
77269-160 77269-162 77269-164
trans-4-(Cyclopropylmethoxy)cyclohexanamine 97%
trans-4-(Cyclopropylmethoxy)cyclohexanamine

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO
MW: 169.26 g/mol
MDL Number: MFCD24644648
CAS Number: 919799-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR