3-Formyl-2,6-dimethoxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dimethoxynicotinaldehyde, 2,6-Dimethoxypyridine-3-carboxaldehyde

A988822-1G A988822-5G
77245-022EA 41.2 USD
77245-022 77245-024
3-Formyl-2,6-dimethoxypyridine 98%
3-Formyl-2,6-dimethoxypyridine

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.17 g/mol
MDL Number: MFCD08064048
CAS Number: 58819-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR