1,10-Phenanthroline-2,9-dicarbaldehyde 95%

Supplier: Ambeed
Warning

A987883-100MG A987883-5G A987883-1G A987883-250MG
77245-018EA 16 USD
77245-018 77245-020 77552-824 77747-738
1,10-Phenanthroline-2,9-dicarbaldehyde 95%
1,10-Phenanthroline-2,9-dicarbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₈N₂O₂
MW: 236.22 g/mol
MDL Number: MFCD00774291
CAS Number: 57709-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR