Chloro{1,3-bis[2,6-bis(1-methylethyl)phenyl]-4,5-dihydroimidazol-2-ylidene}gold(I) 95%

Supplier: Ambeed
Warning

A986322-1G A986322-250MG
77256-978EA 295 USD
77256-978 77256-980
Chloro{1,3-bis[2,6-bis(1-methylethyl)phenyl]-4,5-dihydroimidazol-2-ylidene}gold(I) 95%
Chloro{1,3-bis[2,6-bis(1-methylethyl)phenyl]-4,5-dihydroimidazol-2-ylidene}gold(I)

Precaución: for Research Use Only

Formula: C₂₇H₃₈AuClN₂
MW: 623.02 g/mol
CAS Number: 852445-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR