BRD-73954 98%

Supplier: Ambeed
Warning

Synonyms: BRD73954, N1-Hydroxy-N3-phenethylisophthalamide

A985320-100MG A985320-10MG A985320-1MG A985320-25MG A985320-50MG A985320-5MG
77245-006EA 48 USD
77245-006 77245-008 77245-010 77245-012 77245-014 77245-016
BRD-73954 98%
BRD-73954

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₂O₃
MW: 284.31 g/mol
MDL Number: MFCD26383435
CAS Number: 1440209-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR