(R)-3,3'-Bis(1-naphthalenyl)-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate ≥98%, ee 99%

Supplier: Ambeed
Warning

A984858-100MG A984858-1G A984858-250MG
77256-972EA 50 USD
77256-972 77256-974 77256-976
(R)-3,3'-Bis(1-naphthalenyl)-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate ≥98%, ee 99%
(R)-3,3'-Bis(1-naphthalenyl)-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate

Precaución: for Research Use Only

Formula: C₄₀H₂₅O₄P
CAS Number: 864943-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR