1-Methyl-4-phenylpyridin-1-ium iodide 97%

Supplier: Ambeed
Danger

A984855-100MG A984855-250MG A984855-1G
77256-968EA 82 USD
77256-968 77256-970 77747-736
1-Methyl-4-phenylpyridin-1-ium iodide 97%
1-Methyl-4-phenylpyridin-1-ium iodide

Precaución: for Research Use Only

Formula: C₁₂H₁₂IN
MDL Number: MFCD00055096
CAS Number: 36913-39-0
UN: 2811
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR