(±)-3-(Diethylamino)-1,2-propanediol 98%

Supplier: Ambeed
Warning

Synonyms: (±)-3-(Diethylamino)-1,2-dihydroxy propane, 3-(Diethylamino)propane-1,2-diol

A984681-100G A984681-25G A984681-500G
77282-112EA 29.87 USD
77282-112 77282-114 77282-116
(±)-3-(Diethylamino)-1,2-propanediol 98%
(±)-3-(Diethylamino)-1,2-propanediol

Precaución: for Research Use Only

Formula: C₇H₁₇NO₂
MW: 147.22 g/mol
MDL Number: MFCD00004716
CAS Number: 621-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR