4-(Trifluoromethoxy)anisole 98%

Supplier: Ambeed
Danger

A984225-10G A984225-1G A984225-25G A984225-5G
77294-390EA 11 USD
77294-390 77294-392 77294-394 77294-396
4-(Trifluoromethoxy)anisole 98%
4-(Trifluoromethoxy)anisole

Precaución: for Research Use Only

Formula: C₈H₇F₃O₂
MDL Number: MFCD00216942
CAS Number: 710-18-9
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR