2-Chloro-5-methyl-1H-benzo[d]imidazole 95%

Supplier: Ambeed
Warning

A983444-100MG A983444-1G A983444-250MG
77244-998EA 35.02 USD
77244-998 77595-578 77595-580
2-Chloro-5-methyl-1H-benzo[d]imidazole 95%
2-Chloro-5-methyl-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₈H₇ClN₂
MW: 166.61 g/mol
MDL Number: MFCD11046672
CAS Number: 4887-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR