2-(2-Bromoacetyl)thiophene 95%

Supplier: Ambeed
Danger

Synonyms: 2-Bromo-1-(thiophen-2-yl)ethanone, 2-(Bromoacetyl)thiophene

A981933-250MG A981933-5G
77244-994EA 10.3 USD
77244-994 77618-698
2-(2-Bromoacetyl)thiophene 95%
2-(2-Bromoacetyl)thiophene

Precaución: for Research Use Only

Formula: C₆H₅BrOS
MW: 205.07 g/mol
MDL Number: MFCD02677721
CAS Number: 10531-41-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR