1-Hexadecanol (cetyl alcohol) 95%

Supplier: Ambeed
Warning

Synonyms: Cetyl alcohol, Palmityl alcohol

A981801-100G A981801-500G
77244-990EA 6.18 USD
77244-990 77244-992
1-Hexadecanol (cetyl alcohol) 95%
1-Hexadecanol

Precaución: for Research Use Only

Formula: H₃C(CH₂)₁₄CH₂OH
MW: 242.45 g/mol
MDL Number: MFCD00004760
CAS Number: 36653-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR