Pentifylline 98%

Supplier: Ambeed
Warning

Synonyms: 1-Hexyl-3,7-dimethyl-1H-purine-2,6(3H,7H)-dione, 1-Hexyl-3,7-dimethylxanthine, 3,7-Dimethyl-1-hexylxanthine, 1-Hexyltheobromine, 1-n-Hexyltheobromine, 2,6-Dihydroxy-3,7-dimethyl-1-hexylpurine

A981200-100MG A981200-250MG
77244-986EA 28 USD
77244-986 77244-988
Pentifylline 98%
Pentifylline

Precaución: for Research Use Only

Formula: C₁₃H₂₀N₄O₂
MDL Number: MFCD00041424
CAS Number: 1028-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR