AdBrettPhos Pd G3 98%

Supplier: Ambeed
Warning

A980921-100MG A980921-1G A980921-250MG A980921-5G
77256-952EA 16 USD
77256-952 77256-954 77256-956 77256-958
AdBrettPhos Pd G3 98%
AdBrettPhos Pd G3

Precaución: for Research Use Only

Formula: C₅₆H₇₄NO₅PPdS
MDL Number: MFCD27952546
CAS Number: 1445972-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR