4-Fluoro-3-iodoaniline 97%

Supplier: Ambeed
Warning

A980614-100MG A980614-250MG A980614-1G
77256-948EA 15 USD
77256-948 77256-950 77573-930
4-Fluoro-3-iodoaniline 97%
4-Fluoro-3-iodoaniline
Formula: C₆H₅FIN
MDL Number: MFCD18396914
CAS Number: 647025-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR