Tetrahydro-2H-pyran-4-yl carbonochloridate 96%

Supplier: Ambeed
Danger

A980590-250MG A980590-1G
77269-144EA 214.24 USD
77269-144 77552-810
Tetrahydro-2H-pyran-4-yl carbonochloridate 96%
Tetrahydro-2H-pyran-4-yl carbonochloridate

Precaución: for Research Use Only

Formula: C₆H₉ClO₃
MW: 164.58 g/mol
MDL Number: MFCD20485277
CAS Number: 89641-80-5
UN: 3277
ADR: 6.1,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR