2-Chloro-3,5-dimethylpyrazine 97%

Supplier: Ambeed
Warning

A979422-1G A979422-250MG A979422-5G
77269-138EA 33.99 USD
77269-138 77269-140 77269-142
2-Chloro-3,5-dimethylpyrazine 97%
2-Chloro-3,5-dimethylpyrazine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MW: 142.5900 g/mol
MDL Number: MFCD00126945
CAS Number: 38557-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR