3-Acetyl-4-hydroxy-2H-chromen-2-one 95%

Supplier: Ambeed
Warning

A978964-100MG A978964-1G A978964-250MG A978964-5G
77282-102EA 46.35 USD
77282-102 77282-104 77282-106 77282-108
3-Acetyl-4-hydroxy-2H-chromen-2-one 95%
3-Acetyl-4-hydroxy-2H-chromen-2-one

Precaución: for Research Use Only

Formula: C₁₁H₈O₄
MDL Number: MFCD00138133
CAS Number: 2555-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR