(±)-2-Ethyl-1-hexanol 99.5%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Ethylhexan-1-ol
A978304-100G
A978304-25G
A978304-500G
77269-132EA
7
USD
77269-132
77269-134
77269-136
(±)-2-Ethyl-1-hexanol 99.5%
(±)-2-Ethyl-1-hexanol
Precaución: for Research Use Only
Formula:
CH₃(CH₂)₃CH(C₂H₅)CH₂OH MW: 130.23 g/mol Boiling Pt: 183 Melting Pt: -76 Flash Pt: 81 |
MDL Number:
MFCD00004746 CAS Number: 104-76-7 Merck Index: 13,03842 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...