3-Hydroxypropionic acid ∼ 28-32% in water

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxypropionic Acid, 3-Hydroxypropionic Acid (contains varying amounts of 3,3-Oxydipropionic Acid), Hydracrylic Acid

A977963-100G A977963-100MG A977963-10G A977963-1G A977963-250MG A977963-25G A977963-5G
77282-088EA 805.46 USD
77282-088 77282-090 77282-092 77282-094 77282-096 77282-098 77282-100
3-Hydroxypropionic acid ∼ 28-32% in water
3-Hydroxypropionic acid

Precaución: for Research Use Only

Formula: C₃H₆O₃
MW: 90.08 g/mol
MDL Number: MFCD00058998
CAS Number: 503-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR