3,5-Difluorocinnamic acid 97%

Supplier: Ambeed
Warning

A977858-100G A977858-25G A977858-5G
77269-126EA 215 USD
77269-126 77269-128 77269-130
3,5-Difluorocinnamic acid 97%
3,5-Difluorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₆F₂O₂
MDL Number: MFCD00010321
CAS Number: 84315-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR