(12aR)-1,12-Bis(diphenylphosphino)-6,7-dihydrodibenzo[e,g][1,4]dioxocine 98%

Supplier: Ambeed
Warning

A976565-1G A976565-250MG
77269-122EA 849.75 USD
77269-122 77269-124
(12aR)-1,12-Bis(diphenylphosphino)-6,7-dihydrodibenzo[e,g][1,4]dioxocine 98%
(12aR)-1,12-Bis(diphenylphosphino)-6,7-dihydrodibenzo[e,g][1,4]dioxocine

Precaución: for Research Use Only

Formula: C₃₈H₃₀O₂P₂
MW: 580.59 g/mol
CAS Number: 301847-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR