2,4-Dimethoxy-5-pyrimidineboronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dimethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2,4-Dimethoxypyrimidine-5-boronic acid pinacol ester

A976308-1G A976308-250MG A976308-5G
77256-932EA 58.71 USD
77256-932 77256-934 77256-936
2,4-Dimethoxy-5-pyrimidineboronic acid pinacol ester 96%
2,4-Dimethoxy-5-pyrimidineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₉BN₂O₄
MW: 266.1013 g/mol
MDL Number: MFCD06795654
CAS Number: 936250-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR