4-Aminotetrahydro-2H-pyran-4-carboxylic acid 96%

Supplier: Ambeed
Danger

Synonyms: 4-Aminotetrahydropyran-4-carboxylic Acid

A975709-10G A975709-1G A975709-25G
77294-384EA 123.6 USD
77294-384 77294-386 77294-388
4-Aminotetrahydro-2H-pyran-4-carboxylic acid 96%
4-Aminotetrahydro-2H-pyran-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₁₁NO₃
MW: 145.16 g/mol
MDL Number: MFCD06656112
CAS Number: 39124-20-4
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR