5-Chloro-2-phenyl-1H-indole 95%

Supplier: Ambeed
Warning

A975250-250MG A975250-1G
77244-980EA 77 USD
77244-980 77747-733
5-Chloro-2-phenyl-1H-indole 95%

Precaución: for Research Use Only

Formula: C14H10ClN
MDL Number: MFCD00034706
CAS Number: 23746-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR