(2R)-1-[(4S)-4-(1,1-Dimethylethyl)-4,5-dihydro-2-oxazolyl]-2-(diphenylphosphino)ferrocene ≥98%, ee 99%

Supplier: Ambeed
Warning

A974534-100MG A974534-1G A974534-250MG
77256-924EA 35 USD
77256-924 77256-926 77256-928
(2R)-1-[(4S)-4-(1,1-Dimethylethyl)-4,5-dihydro-2-oxazolyl]-2-(diphenylphosphino)ferrocene ≥98%, ee 99%
(2R)-1-[(4S)-4-(1,1-Dimethylethyl)-4,5-dihydro-2-oxazolyl]-2-(diphenylphosphino)ferrocene

Precaución: for Research Use Only

Formula: C₂₉H₃₀FeNOP
MW: 495.37 g/mol
MDL Number: MFCD19105639
CAS Number: 163169-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR