[1,1':4',1'':4'',1'''-Quaterphenyl]-4,4'''-dicarboxylic acid 98%

Supplier: Ambeed
Warning

A974340-100MG A974340-250MG A974340-1G
77256-920EA 39.14 USD
77256-920 77256-922 77618-678
[1,1':4',1'':4'',1'''-Quaterphenyl]-4,4'''-dicarboxylic acid 98%
[1,1':4',1'':4'',1'''-Quaterphenyl]-4,4'''-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₂₆H₁₈O₄
MW: 394.41 g/mol
MDL Number: MFCD30534208
CAS Number: 143613-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR