GF-627368X 98+%

Supplier: Ambeed
Warning

Synonyms: 4-(4-,9-Diethoxy-1,3-dihydro-1-oxo-2H-benz[f]isoindol-2-yl)-N-(phenylsulphonyl)benzeneacetamide, GW627368, 2-(4-(4,9-Diethoxy-1-oxo-1H-benzo[f]isoindol-2(3H)-yl)phenyl)-N-(phenylsulphonyl)acetamide

A974288-10MG A974288-1MG A974288-25MG A974288-5MG
77244-968EA 71.07 USD
77244-968 77244-970 77244-972 77244-974
GF-627368X 98+%
GF-627368X

Precaución: for Research Use Only

Formula: C₃₀H₂₈N₂O₆S
MW: 544.62 g/mol
MDL Number: MFCD12912327
CAS Number: 439288-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR