4-Chloro-2,5-dimethoxyaniline 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dimethoxy-4-chloroaniline

A973014-100G A973014-25G A973014-500G
77294-376EA 23.69 USD
77294-376 77294-378 77294-380
4-Chloro-2,5-dimethoxyaniline 98%
4-Chloro-2,5-dimethoxyaniline

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO₂
MW: 187.63 g/mol
MDL Number: MFCD00014893
CAS Number: 6358-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR