2-Chloro-6-methoxybenzonitrile 98%

Supplier: Ambeed
Warning

A972751-10G A972751-1G A972751-25G
77269-104EA 72 USD
77269-104 77269-106 77269-108
2-Chloro-6-methoxybenzonitrile 98%
2-Chloro-6-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆ClNO
MDL Number: MFCD00019743
CAS Number: 6575-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR