3-[[3,5-Bis(trifluoromethyl)phenyl]amino]-4-[[(8α,9S)-10,11-dihydro-6'-methoxycinchonan-9-yl]amino]-3-cyclobutene-1,2-dione 98%

Supplier: Ambeed
Warning

A972154-1G A972154-250MG
77256-908EA 592.25 USD
77256-908 77256-910
3-[[3,5-Bis(trifluoromethyl)phenyl]amino]-4-[[(8α,9S)-10,11-dihydro-6'-methoxycinchonan-9-yl]amino]-3-cyclobutene-1,2-dione 98%
3-[[3,5-Bis(trifluoromethyl)phenyl]amino]-4-[[(8α,9S)-10,11-dihydro-6'-methoxycinchonan-9-yl]amino]-3-cyclobutene-1,2-dione

Precaución: for Research Use Only

Formula: C₃₂H₂₈F₆N₄O₃
MW: 630.58 g/mol
MDL Number: MFCD31707562
CAS Number: 1352957-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR