Fmoc-β-(2-quinolyl)-Ala-OH 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-β-(2-quinolyl)-Ala-OH

A971425-100MG A971425-250MG
77282-084EA 48 USD
77282-084 77282-086
Fmoc-β-(2-quinolyl)-Ala-OH 97%
Fmoc-beta-(2-quinolyl)-Ala-OH

Precaución: for Research Use Only

Formula: C₂₇H₂₂N₂O₄
MDL Number: MFCD01318745
CAS Number: 214852-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR