Ethyl 1H-1,2,3-benzotriazole-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1H-benzo[d][1,2,3]triazole-5-carboxylate

A970836-250MG A970836-5G
77244-964EA 36 USD
77244-964 77244-966
Ethyl 1H-1,2,3-benzotriazole-5-carboxylate 98%
Ethyl 1H-1,2,3-benzotriazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₉H₉N₃O₂
MDL Number: MFCD00963122
CAS Number: 73605-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR