2,6-Dibromo-4-fluorophenol 95%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dibromo-5-fluoro-2-hydroxybenzene

A969321-1G A969321-5G
77294-372EA 6 USD
77294-372 77294-374
2,6-Dibromo-4-fluorophenol 95%
2,6-Dibromo-4-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₃Br₂FO
MDL Number: MFCD00002317
CAS Number: 344-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR