RBC8 98+%

Supplier: Ambeed
Warning

A968704-10MG A968704-1MG A968704-50MG A968704-5MG
77244-956EA 36 USD
77244-956 77244-958 77244-960 77244-962
RBC8 98+%
RBC8

Precaución: for Research Use Only

Formula: C₂₅H₂₀N₄O₃
CAS Number: 361185-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR