2-Diphenylphosphino-2',6'-bis(dimethylamino)-1,1'-biphenyl 98%

Supplier: Ambeed
Warning

A968407-100MG A968407-1G A968407-250MG
77256-894EA 67.98 USD
77256-894 77256-896 77256-898
2-Diphenylphosphino-2',6'-bis(dimethylamino)-1,1'-biphenyl 98%
2-Diphenylphosphino-2',6'-bis(dimethylamino)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₂₈H₂₉N₂P
MW: 424.51 g/mol
MDL Number: MFCD29059466
CAS Number: 1447963-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR