2-Amino-6-ethylbenzoic acid 98%

Supplier: Ambeed
Warning

A968291-100MG A968291-1G A968291-250MG A968291-25G A968291-5G
77269-094EA 19.57 USD
77269-094 77269-096 77269-098 77269-100 77269-102
2-Amino-6-ethylbenzoic acid 98%
2-Amino-6-ethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.18 g/mol
MDL Number: MFCD11616258
CAS Number: 66232-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR