2-Methyl-4-trifluoromethylpyrimidine-5-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Methyl-4-(trifluoromethyl)pyrimidine-5-carboxylic acid
A966854-100MG
A966854-1G
A966854-250MG
77282-070EA
51.5
USD
77282-070
77282-072
77282-074
2-Methyl-4-trifluoromethylpyrimidine-5-carboxylic acid 95%
2-Methyl-4-trifluoromethylpyrimidine-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₅F₃N₂O₂ MW: 206.13 g/mol |
MDL Number:
MFCD06409005 CAS Number: 149771-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...